Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-5-chloro-1,2,4-thiadiazole 97%

Supplier: Thermo Scientific Chemicals
Warning

29791-0100
TS29791-0100EA 377 USD InStock
TS29791-0100
3-Bromo-5-chloro-1,2,4-thiadiazole 97%
3-Bromo-5-chloro-1,2,4-thiadiazole


Formula: C₂BrClN₂S
MW: 199.45 g/mol
Boiling Pt: 192…194 °C (760 mmHg)
Melting Pt: 24…25 °C
Storage Temperature: Ambient
MDL Number: MFCD00799287
CAS Number: 37159-60-7
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5965 to 1.5985 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty