Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Chromotropic acid disodium salt dihydrate ACS

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4,5-Dihydroxy-2,7-naphthalenedisulfonic acid disodium salt dihydrateDisodium 4,5-dihydroxynaphthalene-2,7-disulphonate dihydrate4,5-Dihydroxynaphthalene-2,7-disulphonic acid disodium salt dihydrate4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrateDisodium 4,5-dihydroxynaphthalene-2,7-disulfonate dihydrate4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate4,5-Dihydroxynaphthalene-2,7-disulphonic acid, disodium salt dihydrate4,5-Dihydroxy-2,7-naphthalenedisulphonic acid disodium salt dihydrate

30060-1000 30060-0250 30060-0100
TS30060-0250EA 114.67 USD InStock
TS30060-0250 TS30060-1000 TS30060-0100
Chromotropic acid disodium salt dihydrate ACS
Chromotropic acid disodium salt dihydrate


Formula: (HO)₂C₁₀H₄(SO₃Na)₂·2H₂O
MW: 400.29 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150612
CAS Number: 5808-22-0
EINECS: 204-972-9
Order Now
Specification Test Results
Appearance Off-white to beige to grey to light brown Powder
Infrared spectrum Conforms
Sensitivity (to nitrate) Passes test
Sensitivity (to formaldehyde) Passes test
Clarity of solution Passes test

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty