Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromobutyryl chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

30344-0250 30344-0050
TS30344-0250EA 149.2 USD InStock
TS30344-0250 TS30344-0050
4-Bromobutyryl chloride 95%
4-Bromobutyryl chloride


Formula: C₄H₆BrClO
Boiling Pt: 101 °C (37 mmHg)
Density: 1.602 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00000753
CAS Number: 927-58-2
EINECS: 213-154-0
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow to light pink Liquid
Infrared spectrum Conforms
Refractive index 1.4910 to 1.4940 (20°C, 589 nm)
GC ≥94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty