Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Ethylbenzoyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Ethylbenzene-1-carbonyl chloride

30573-0050 30573-0250
TS30573-0050EA 62 USD InStock
TS30573-0050 TS30573-0250
4-Ethylbenzoyl chloride 97%
4-Ethylbenzoyl chloride


Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 235…236 °C (760 mmHg)
Melting Pt: 42…43 °C
Density: 1.14 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000697
CAS Number: 16331-45-6
EINECS: 240-404-6
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5465 to 1.5485 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty