Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chromotropic acid disodium salt dihydrate

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4,5-Dihydroxy-2,7-naphthalenedisulfonic acid disodium salt dihydrateDisodium 4,5-dihydroxynaphthalene-2,7-disulphonate dihydrate4,5-Dihydroxynaphthalene-2,7-disulphonic acid disodium salt dihydrate4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrateDisodium 4,5-dihydroxynaphthalene-2,7-disulfonate dihydrate4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate4,5-Dihydroxynaphthalene-2,7-disulphonic acid, disodium salt dihydrate4,5-Dihydroxy-2,7-naphthalenedisulphonic acid disodium salt dihydrate

37810-2500
TS37810-2500EA 183.31 USD InStock
TS37810-2500
Chromotropic acid disodium salt dihydrate
Chromotropic acid disodium salt dihydrate


Formula: (HO)₂C₁₀H₄(SO₃Na)₂·2H₂O
MW: 400.29 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150612
CAS Number: 5808-22-0
EINECS: 204-972-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty