Would you like to log in again?
Envy® Liquid Disinfectant Cleaner
Supplier: Diversey
04528
76547-676CT 155.14 USD InStock
76547-676
Envy® Liquid Disinfectant Cleaner
Cleaning Agents Disinfectants
A quaternary-based disinfectant and virucidal cleaner in RTU format.
  • Ready-to-use liquid disinfects in five minutes and is a non-food contact sanitizer in 30 seconds. It is highly effective against a wide variety of pathogenic microorganisms including bacteria, antibiotic resistant bacteria, viruses, fungi and mildew
  • Cleans and disinfects surfaces. Bactericidal, fungicidal, mildewstatic and virucidal
  • Quickly removes dirt, grime, mold, mildew and other common soils on hard, nonporous surfaces. Clean lavender fragrance
The RTU liquid disinfects in five minutes and is a non-food contact sanitizer in 30 seconds.
Colorless with a lavender scent.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty