Would you like to log in again?
Dilac® EG, Heavy Duty Low Foaming Acid Detergent
Supplier: Diversey
Dilac®
57552010
76547-728EA 2546.07 USD InStock
76547-728
Dilac® EG, Heavy Duty Low Foaming Acid Detergent
Cleaning Agents Detergents and Surface Cleansers
Dilac® EG cleaner is a heavy duty, phosphoric acid-based liquid detergent, rust remover and descaler for use in a wide range of applications across all food and beverage applications.
  • Highly acidic and effective at removing rust, inorganic soils, proteins and starch
  • Suitable for clean-in-place (CIP), clean-out-of-place (COP) and general spray washing applications
  • Conductive liquid detergent suitable for automatic dosing and control
  • Conductive liquid detergent suitable for automatic dosing and control
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty