Would you like to log in again?
Chek-Stix® UA Diptube Liquid QC, Siemens Healthineers
Supplier: Siemens Healthineers
Chek-Stix®
11561694
76554-124KT 225.86 USD InStock
76554-124
Chek-Stix® UA Diptube Liquid QC, Siemens Healthineers
Clinical Diagnostic Supplies Clinical Reagents and Kits
Chek-Stix UA Diptube Liquid QC contains both Level 1 (negative for all chemistry parameters and positive for hCG) and Level 2 (positive for all chemistry parameters and negative for hCG), packaged in one box.
  • Liquid-ready solution means less time prepping quality control solution and more time running patient samples
  • Barcode-enables for quick, accurate data input
  • Supports our broad urine chemistry and sediment parameter menu
  • Validated for use with all Siemens Healthineers urinalysis devices and reagents
  • 22 month's shelf life from manufacture date
Both Level 1 and Level 2 diptube controls are ready-to-use liquid quality controls for use with the CLINITEK Status®+ Analyzer, CLINITEK Advantus® Analyzer, and visual-read urine strips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty