Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-4-nitrobenzene 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Chloronitrobenzene

10963-0050 10963-0010 10963-1000
TS10963-0010EA 97 USD InStock
TS10963-0010 TS10963-1000 TS10963-0050
1-Chloro-4-nitrobenzene 99%
1-Chloro-4-nitrobenzene


Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Boiling Pt: 239 °C (1013 hPa)
Melting Pt: 81…84 °C
Density: 1.298 g/cm³
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00007285
CAS Number: 100-00-5
EINECS: 202-809-6
UN: 1578
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Yellow Crystals or flakes
Infrared spectrum Conforms
Melting point 81 to 84 °C
GC ≥98.5 %
Titration with HClO4 98.5 to 101.5 % (on dried substance)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty