Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Fluoro-4-nitrobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Fluoronitrobenzene1-Fluoro-4-nitrobenzene

11949-1000 11949-5000
TS11949-1000EA 66 USD InStock
TS11949-1000 TS11949-5000
1-Fluoro-4-nitrobenzene 99%
1-Fluoro-4-nitrobenzene


Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 27 °C
Density: 1.33 g/cm³
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00007282
CAS Number: 350-46-9
EINECS: 206-502-8
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear yellow to dark brown Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5302 to 1.5322 (20 °C, 589 nm)
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty