Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-5-bromovalerate 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 5-Bromovaleric acid methyl esterMethyl 5-Bromovalerate

12646-0100 12646-0250
TS12646-0100EA 85 USD InStock
TS12646-0100 TS12646-0250
Methyl-5-bromovalerate 97%
Methyl 5-bromovalerate


Formula: BrH₂C(CH₂)₃COOCH₃
MW: 195.06 g/mol
Boiling Pt: 79…80 °C (5.33 hPa)
Density: 1.36 g/cm³
Flash Pt: 99 °C
MDL Number: MFCD00000265
CAS Number: 5454-83-1
EINECS: 226-709-7
Order Now
Specification Test Results
Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4620 to 1.4640 (20 °C, 589 nm)
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty