Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl sulfone 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: DimethylsulfoneMethyl sulfoneMethyl sulphoneDimethylsulphone

12778-0025 12778-5000 12778-0050
TS12778-0050EA 24.67 USD InStock
TS12778-0050 TS12778-5000 TS12778-0025
Dimethyl sulfone 98%
Dimethyl sulphone


Formula: C₂H₆O₂S
MW: 94.13 g/mol
Boiling Pt: 238 °C (760 mmHg)
Melting Pt: 107…111 °C
Flash Pt: 143 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007566
CAS Number: 67-71-0
EINECS: 200-665-9
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 107 to 111 °C
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty