Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitrobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: o-Nitrobenzonitrile

12847-0100
TS12847-0100EA 52 USD InStock
TS12847-0100
2-Nitrobenzonitrile 98%
2-Nitrobenzonitrile


Formula: C₇H₄N₂O₂
MW: 148.12 g/mol
Boiling Pt: 165 °C (16 mmHg)
Melting Pt: 110.8…111.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00007044
CAS Number: 612-24-8
EINECS: 210-301-0
UN: 3439
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 107 to 112 °C
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty