Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ω-Bromo-4-chloroacetophenone 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Bromo-4′-chloroacetophenone2-Bromo-1-(4-chlorophenyl)ethanone4-Chlorophenacyl bromide2-Bromo-4'-chloroacetophenoneBromomethyl p-chlorophenyl ketoneα-Bromo-4-chloroacetophenone

14636-0250 14636-1000
TS14636-0250EA 45 USD InStock
TS14636-0250 TS14636-1000
ω-Bromo-4-chloroacetophenone 98+%
ω-Bromo-4-chloroacetophenone


Formula: C₈H₆BrClO
MW: 233.49 g/mol
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 95…99 °C
MDL Number: MFCD00000625
CAS Number: 536-38-9
EINECS: 208-631-5
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance White to light yellow Crystals
Infrared spectrum Conforms
Melting point 95 to 99 °C
Loss on drying ≤1 % (70 °C, 2 h)
GC ≥98.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty