Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopentylmethanol 98%

Supplier: Thermo Scientific Chemicals

14715-0100
TS14715-0100EA 142 USD InStock
TS14715-0100
Cyclopentylmethanol 98%
Cyclopentylmethanol


Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 162 °C (1013 hPa)
Density: 0.92 g/cm³
Flash Pt: 62 °C
MDL Number: MFCD00001384
CAS Number: 3637-61-4
EINECS: 222-861-3
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4570 to 1.4590 (20 °C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty