Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,1,3,3-Tetramethoxypropane 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Malonaldehyde bis(dimethyl acetal)

14861-5000 14861-1000 14861-0051
TS14861-1000EA 36 USD InStock
TS14861-1000 TS14861-0051 TS14861-5000
1,1,3,3-Tetramethoxypropane 98%
1,1,3,3-Tetramethoxypropane


Formula: C₇H₁₆O₄
MW: 164.2 g/mol
Boiling Pt: 183 °C (760 mmHg)
Density: 0.99 g/cm³
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00008488
CAS Number: 102-52-3
EINECS: 203-037-2
UN: 3271
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4050 to 1.4090 (20 °C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty