Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Formylbenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-CarboxybenzaldehydePhthalaldehydic acid

15208-0250 15208-1000
TS15208-1000EA 111 USD InStock
TS15208-1000 TS15208-0250
2-Formylbenzoic acid 99%
2-Formylbenzoic acid


Formula: C₈H₆O₃
MW: 150.13 g/mol
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00003336
CAS Number: 119-67-5
EINECS: 204-342-3
Order Now
Specification Test Results
Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 96 to 99 °C
HPLC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty