Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dibromosalicylaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3,5-Dibromo-2-hydroxybenzaldehyde

15340-1000 15340-0010 15340-0250
TS15340-1000EA 345 USD InStock
TS15340-1000 TS15340-0010 TS15340-0250
3,5-Dibromosalicylaldehyde 98%
3,5-Dibromosalicylaldehyde


Formula: C₇H₄Br₂O₂
MW: 279.92 g/mol
Melting Pt: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00003318
CAS Number: 90-59-5
EINECS: 202-003-4
Order Now
Specification Test Results
Appearance Yellow to yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 80 to 84 °C
Titration with NaOH ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty