Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Caprinaldehyde 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: DecylaldehydeDecyl aldehydeDecanal

15497-1000 15497-5000
TS15497-1000EA 32.6 USD InStock
TS15497-1000 TS15497-5000
Caprinaldehyde 95%
Caprinaldehyde


Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 207…209 °C (760 mmHg)
Melting Pt: 7 °C
Density: 0.83 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007031
CAS Number: 112-31-2
EINECS: 203-957-4
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4270 to 1.4320 (20 °C, 589 nm)
GC ≥94 %
NMR (1H-NMR) Consistent with structure

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty