Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Nonanol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: n-NonanolOctyl carbinol1-Hydroxy nonanen-Nonyl alcoholNonan-1-ol1-Nonyl alcoholNonyl alcohol

15747-1000 15747-2500 15747-0010
TS15747-2500EA 63 USD InStock
TS15747-2500 TS15747-0010 TS15747-1000
1-Nonanol 97%
1-Nonanol


Formula: H₃C(CH₂)₇CH₂OH
MW: 144.26 g/mol
Boiling Pt: 213…215 °C (760 mmHg)
Melting Pt: –5 °C
Density: 0.827 g/cm³
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00002990
CAS Number: 143-08-8
EINECS: 205-583-7
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4324 to 1.4344 (20 °C, 589 nm)
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty