Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethoxyacetic acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Ethoxyacetic acid

15986-1000 15986-0250
TS15986-0250EA 203 USD InStock
TS15986-0250 TS15986-1000
Ethoxyacetic acid 98%
Ethoxyacetic acid


Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 97…100 °C (11 mmHg)
Density: 1.102 g/cm³
Flash Pt: 97 °C
MDL Number: MFCD00004310
CAS Number: 627-03-2
EINECS: 210-978-2
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4200 (20 °C, 589 nm)
GC ≥97.5 %
Residue after evaporation ≤0.0005 %
Acidity/alkalinity ≤0.0005 meq/g
Filt.over 0.2 micron PFTE Filtered

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty