Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Phenyl-1-propanol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Phenyl-1-Propanol

16131-0010 16131-2500
TS16131-2500EA 48 USD InStock
TS16131-2500 TS16131-0010
3-Phenyl-1-propanol 98%
3-Phenyl-1-propanol


Formula: (C₆H₅)CH₂CH₂CH₂OH
MW: 136.19 g/mol
Boiling Pt: 455 °C (1013 hPa)
Melting Pt: –18 °C
Density: 1.04 g/cm³
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00002950
CAS Number: 122-97-4
EINECS: 204-587-6
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5270 (20 °C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty