Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isovaleraldehyde 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Methylbutyraldehyde

16397-0025 16397-1000 16397-5000
TS16397-5000EA 81 USD InStock
TS16397-5000 TS16397-1000 TS16397-0025
Isovaleraldehyde 98%
Isovaleraldehyde


Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 90 °C (760 mmHg)
Melting Pt: –28 °C
Density: 0.796 g/cm³
Flash Pt: –5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007014
CAS Number: 590-86-3
EINECS: 209-691-5
UN: 2058
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3870 to 1.3890 (20 °C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty