Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Dichloroacetone 99%

Supplier: Thermo Scientific Chemicals
Danger

17363-0500 17363-2500 17363-0250
TS17363-0500EA 79 USD InStock
TS17363-0500 TS17363-2500 TS17363-0250
1,3-Dichloroacetone 99%
1,3-Dichloroacetone


Formula: ClH₂CC(O)CH₂Cl
MW: 126.97 g/mol
Boiling Pt: 173 °C (1013 hPa)
Melting Pt: 38…42 °C
Density: 1.383 g/cm³
Flash Pt: 89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000937
CAS Number: 534-07-6
EINECS: 208-585-6
UN: 2649
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Colorless to white Crystals
Infrared spectrum Conforms
Melting point 38 to 42 °C
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty