Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyruvaldehyde (methylglyoxal) 30-45 (w/w) in water

Supplier: Thermo Scientific Chemicals
Danger

17579-0025 17579-5000 17579-1000
TS17579-0025EA 1679 USD InStock
TS17579-0025 TS17579-1000 TS17579-5000
Pyruvaldehyde (methylglyoxal) 30-45 (w/w) in water
Pyruvaldehyde (Methylglyoxal)


Formula: C₃H₄O₂
Boiling Pt: 72 °C
Melting Pt: –20 °C
Density: 1.2 g/cm³
Flash Pt: >71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006960
CAS Number: 78-98-8
EINECS: 201-164-8
Order Now
Specification Test Results
Appearance Clear yellow to brown Solution
Refractive index 1.3910 to 1.4200 (20°C, 589 nm)
Titration with NaOH 30 to 45 %
HPLC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty