Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xanthotoxin 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 9-Methoxy-7H-furo[3,2-g]chromen-7-oneMethoxsalen8-Methoxypsoralen

21415-0050 21415-0010
TS21415-0010EA 48 USD InStock
TS21415-0010 TS21415-0050
Xanthotoxin 99%
Xanthotoxin


Formula: C₁₂H₈O₄
MW: 216.19 g/mol
Melting Pt: 143…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00005009
CAS Number: 298-81-7
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance White to cream Powder or crystals or needles
Infrared spectrum Conforms
Melting point 143 to 148 °C
Water ≤0.5 % (K.F.)
HPLC ≥98.5 % (on anhydrous substance)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty