Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium ethyl malonate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Monoethyl malonate potassium saltEthyl potassium malonate

24808-0250 24808-5000 24808-1000
TS24808-0250EA 64.91 USD InStock
TS24808-0250 TS24808-1000 TS24808-5000
Potassium ethyl malonate 98%
Potassium ethyl malonate


Formula: C₅H₇KO₄
MW: 170.21 g/mol
Melting Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00035603
CAS Number: 6148-64-7
EINECS: 228-156-7
Order Now
Specification Test Results
Appearance White to cream Powder or crystals
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty