Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trifluoromethoxy)benzaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

29350-0100 29350-0010
TS29350-0010EA 24 USD InStock
TS29350-0010 TS29350-0100
4-(Trifluoromethoxy)benzaldehyde 99%
4-(Trifluoromethoxy)benzaldehyde


Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 80 °C (16 mmHg)
Density: 1.33 g/cm³
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00041530
CAS Number: 659-28-9
EINECS: 211-531-4
Order Now
Specification Test Results
Appearance Clear colorless to yellow-green Liquid
Infrared spectrum Conforms
Refractive index 1.4570 to 1.4590 (20 °C, 589 nm)
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty