Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2',4'-Dichloroacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

30243-0250 30243-2500
TS30243-2500EA 224 USD InStock
TS30243-2500 TS30243-0250
2',4'-Dichloroacetophenone 97%
2',4'-Dichloroacetophenone


Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 29…34 °C
Density: 1.32 g/cm³
Flash Pt: 131 °C
MDL Number: MFCD00000581
CAS Number: 2234-16-4
EINECS: 218-780-8
Order Now
Specification Test Results
Appearance Colorless to yellow-brown Solid
Infrared spectrum Conforms
Melting point 29 to 33 °C
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty