Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylumbelliferyl-β-D-xylopyranoside 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Methylumbelliferyl-β-D-xylopyranoside, β-xylosidase substrate

33727-2500
TS33727-2500EA 422.16 USD InStock
TS33727-2500
4-Methylumbelliferyl-β-D-xylopyranoside 99%
4-Methylumbelliferyl-beta-D-xylopyranoside


Formula: C₁₅H₁₆O₇
MW: 308.286 g/mol
Melting Pt: 223 °C
Storage Temperature: Freezer
MDL Number: MFCD00037607
CAS Number: 6734-33-4
Order Now
Specification Test Results
Appearance White to almost white Powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
HPLC ≥98.5 %
Appearance of solution (1% in DMF) Almost clear, colorless to light yellow
Specific optical rotation -28° to -32° (20°C, 589 nm) (c=0.3, pyridine)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty