Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzothiazol-2-ylmethanol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(Hydroxymethyl)benzothiazole2-Benzothiazolemethanol2-Hydroxymethylbenzothiazole

35850-0010
TS35850-0010EA 49 USD InStock
TS35850-0010
Benzothiazol-2-ylmethanol 98%
Benzothiazol-2-ylmethanol


Formula: C₈H₇NOS
MW: 165.21 g/mol
Melting Pt: 95…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00226293
CAS Number: 37859-42-0
Order Now
Specification Test Results
Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 98 °C
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty