Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzyl-4-bromophenyl ketone

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-(4-Bromophenyl)-2-phenylethanone

38189-0010 38189-0050
TS38189-0010EA 45.44 USD InStock
TS38189-0010 TS38189-0050
Benzyl-4-bromophenyl ketone
Benzyl 4-bromophenyl ketone


Formula: C₁₄H₁₁BrO
MW: 275.14 g/mol
Melting Pt: 109…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00016331
CAS Number: 2001-29-8
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 110 to 115 °C
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty