Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-2-(1-Hydroxyethyl)pyridine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (R)-1-(Pyridin-2-yl)ethanol

38286-0010 38286-2500
TS38286-0010EA 173 USD InStock
TS38286-0010 TS38286-2500
(R)-2-(1-Hydroxyethyl)pyridine 98%
(R)-2-(1-Hydroxyethyl)pyridine


Formula: C₇H₉NO
MW: 123.155 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04972322
CAS Number: 27911-63-3
Order Now
Specification Test Results
Appearance White to grey Crystals or solid
Infrared spectrum Conforms
GC ≥97.5 %
Specific optical rotation +24° to +31° (20°C, 589 nm) (c=1, toluene)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty