Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Hexynoic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

38370-0050 38370-0250 38370-0010
TS38370-0010EA 57 USD InStock
TS38370-0010 TS38370-0250 TS38370-0050
5-Hexynoic acid 97%
5-Hexynoic acid


Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: 41 °C
Density: 1.016 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00066346
CAS Number: 53293-00-8
UN: 2922
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4480 to 1.4500 (20 °C, 589 nm)
Titration with NaOH ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty