Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-6-formylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Bromopyridine-2-carboxaldehyde6-Bromopyridine-2-carbaldehyde6-Bromo-2-pyridinecarboxaldehyde

38742-0050 38742-0010
TS38742-0050EA 128 USD InStock
TS38742-0050 TS38742-0010
2-Bromo-6-formylpyridine 97%
2-Bromo-6-formylpyridine


Formula: C₆H₄BrNO
MW: 186.01 g/mol
Melting Pt: 79…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683546
CAS Number: 34160-40-2
Order Now
Specification Test Results
Appearance White to beige to light brown Powder
Infrared spectrum Conforms
Melting point 81 to 84 °C
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty