Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Difluoro-2-nitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,6-Difluoronitrobenzene

39734-0050
TS39734-0050EA 238 USD InStock
TS39734-0050
1,3-Difluoro-2-nitrobenzene 98%
1,3-Difluoro-2-nitrobenzene


Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 91…92 °C (11 mmHg)
Density: 1.503 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00192035
CAS Number: 19064-24-5
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear yellow to orange Liquid or low melting solid
Infrared spectrum Conforms
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty