Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Azelaic acid 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 1,9-Nonanedioic acid1,7-Heptanedicarboxylic acid

40152-5000 40152-1000 40152-0250
TS40152-0250EA 27.6 USD InStock
TS40152-0250 TS40152-5000 TS40152-1000
Azelaic acid 98%
Azelaic acid


Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Boiling Pt: 237 °C (20hPa)
Melting Pt: 98…103 °C
Flash Pt: 215 °C
Storage Temperature: Ambient
MDL Number: MFCD00004432
CAS Number: 123-99-9
EINECS: 204-669-1
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty