Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl valerate 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Methyl PentanoateValeric acid methyl ester

41505-0250
TS41505-0250EA 38.77 USD InStock
TS41505-0250
Methyl valerate 99%
Methyl valerate


Formula: C₆H₁₂O₂
Boiling Pt: 126 °C
Melting Pt: –91 °C
Density: 0.89 g/cm³
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00009478
CAS Number: 624-24-8
EINECS: 210-838-0
UN: 3272
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3965 to 1.3985 (20 °C, 589 nm)
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty