Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oxalacetic acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Oxalacetic acid2-Succinic acid

41660-1000 41660-0050 41660-0250
TS41660-1000EA 797 USD InStock
TS41660-1000 TS41660-0050 TS41660-0250
Oxalacetic acid 98%
Oxalacetic acid


Formula: C₄H₄O₅
MW: 132.07 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 161 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00002592
CAS Number: 328-42-7
EINECS: 206-329-8
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty