Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trilaurin (glycerol trilaurate) 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Glycerol tridodecanoateGlyceryl trilaurateGlyceryl tridodecanoateGlycerol trilaurate

42183-0250 42183-0050
TS42183-0250EA 330 USD InStock
TS42183-0250 TS42183-0050
Trilaurin (glycerol trilaurate) 98%
Trilaurin (Glycerol trilaurate)


Formula: C₃₉H₇₄O₆
MW: 639.01 g/mol
Melting Pt: 45…47 °C
Density: 0.9 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00026559
CAS Number: 538-24-9
EINECS: 208-687-0
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
TLC ≥97.5 %
pH 6.0 to 9.2 (5 % at 25°C)
Alkalinity ≤0.022 % (as KOH)
Iodate (IO3) ≤2 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤10 ppm
Nitrogen compounds (as N) ≤10 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty