Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trimyristin 90%

Supplier: Thermo Scientific Chemicals
Synonyms: Propane-1,2,3-Triyl TritetradecanoateGlycerol Trimyristate

42209-0050 42209-0250
TS42209-0050EA 152 USD InStock
TS42209-0050 TS42209-0250
Trimyristin 90%
Trimyristin


Formula: C₄₅H₈₆O₆
MW: 723.18 g/mol
Melting Pt: 54…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00036229
CAS Number: 555-45-3
Order Now
Specification Test Results
Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
TLC ≥88 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty