Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Hexenoic acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 5-Hexenoicacid

44952-0050 44952-0010
TS44952-0050EA 190 USD InStock
TS44952-0050 TS44952-0010
5-Hexenoic acid 98%
5-Hexenoic acid


Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 203 °C (760 mmHg)
Density: 0.961 g/cm³
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00046558
CAS Number: 1577-22-6
UN: 3265
ADR: 8,III
Order Now
Specification Test Results
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4360 (20 °C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty