Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Heptynoic acid 95%

Supplier: Thermo Scientific Chemicals
Danger

45739-0010 45739-0250 45739-0050
TS45739-0050EA 150 USD InStock
TS45739-0050 TS45739-0250 TS45739-0010
6-Heptynoic acid 95%
6-Heptynoic acid


Formula: C₇H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00239430
CAS Number: 30964-00-2
UN: 3265
Order Now
Specification Test Results
Appearance Colorless to yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥94.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty