Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methylcaproic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

45741-1000 45741-0250 45741-0050
TS45741-1000EA 364 USD InStock
TS45741-1000 TS45741-0050 TS45741-0250
5-Methylcaproic acid 98%
5-Methylcaproic acid


Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 121 °C (30 mmHg)
Density: 0.905 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00036494
CAS Number: 628-46-6
UN: 3265
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to amber Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4220 (20 °C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty