Would you like to log in again?
Floating Microtube Racks, Polypropylene
Supplier: United Scientific Supplies
63201
470338-926EA 97.6 USD InStock
470338-926
Floating Microtube Racks, Polypropylene
Racks Tube Racks
Easy to grab polypropylene floating rack.
  • Easy grab handle
  • Alpha Numeric grid
Floating microtube racks are ideal for thawing, cooling, or incubating samples in water or ice baths, at a wide range of temperatures (from −70 to +100 °C). Tubes are supported below the lip while conical ends are immersed in water. Racks feature a handle for easy removal from water bath, as well as a molded-in alpha numeric grid for identification of samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty