Would you like to log in again?
Thermal Conductivity Bars
Supplier: United Scientific Supplies
CBR001
470339-834EA 46.4 USD InStock
470339-834
Thermal Conductivity Bars
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Visually demonstrate the flow of heat in different metals.
  • Four metals: steel, brass, aluminum, and copper
  • Display temperature range: 35 to 42 °C, × 1 °C
  • Liquid crystal thermometer
This simple and ingenious device quickly and graphically shows students how different metals conduct heat at different rates. It consists of four identically sized bars of common metals (steel, brass, aluminum, and copper) mounted parallel to one another in a plastic support. Attached to each bar is a liquid crystal thermometer. When the ends of the bars are immersed in hot water, the thermometers quickly show the progress of heat along each bar. Students can measure the time for the tops of the bars to reach the top display temperature (42 °C) and discover how well each metal conducts heat.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty