Would you like to log in again?
Gas Measuring Tube
Supplier: United Scientific Supplies
GTUBE50
470339-958EA 21.55 USD InStock
470339-958
Gas Measuring Tube
Tubes Gas Sampling Tubes
Graduated borosilicate tube for measuring gas.
  • Borosilicate glass tube with 50 ml capacity
  • Closed at zero end for gas measurement
  • Autoclavable
Borosilicate glass tube features top-down graduations in durable white enamel, at 0.5 ml intervals, from 0.5 to 50 ml. Tube measures 650 mm long and 13 mm in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty