Would you like to log in again?
Economy Inclined Plane
Supplier: United Scientific Supplies
INPM01
470339-944EA 29.85 USD InStock
470339-944
Economy Inclined Plane
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Economical inclined plane with pulley.
  • Economical inclined plane, 80L×12W cm
  • Made of medium-density fiberboard (MDF)
Economical version of the traditional wood inclined plane. Made of medium-density fiberboard (MDF). Includes adjustable-angle pulley and a 14 mm diameter cross hole for support rod.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty