Would you like to log in again?
LaPlace's Law Demonstrator
Supplier: United Scientific Supplies
LLDE01
470338-988EA 47.3 USD InStock
470338-988
LaPlace's Law Demonstrator
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Device designed to teach Fleming’s left hand rule.
  • Includes activity guide
A great device for teaching Fleming’s left hand rule, this demonstrator consists of a base with two separate parallel conductors, a large U-shaped magnet that fits between the parallel conductors, and a conducting rod attached to a plastic disc. When a current is passed through the rods, a force is exerted on the rod with the disc which causes it to roll along the parallel rods. Includes activity guide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty