Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Lung Apparatus
Supplier: United Scientific Supplies
LUNG01-B
470338-880EA 62.5 USD InStock
470338-880
Lung Apparatus
Models Human Biology Models
Demonstrate how the diaphragm causes the lungs to inflate.
  • Balloons fill, representing the lungs
  • Sealed system similar to the human chest cavity
  • Includes activity guide
Demonstrates how the lungs are filled and emptied with air through contraction and relaxation of the diaphragm muscle. The two balloons act as the lungs and the rubber sheet acts as the diaphragm. Pulling the rubber sheet downward creates pressure within the chamber, and causes the balloons to expand. Releasing the rubber sheet deflates the balloons.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty